Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010120.5 | NP_034250.3 | 3 | 472 | ||
GenBank mRNA | AF026481.1 | - | 2 | 1291 | ||
AK009026.1 | - | 3 | 443 | |||
AK013268.1 | - | 3 | 471 | |||
AK019451.1 | - | 3 | 473 | |||
AK150651.1 | - | 3 | 469 | |||
AK150673.1 | - | 3 | 445 | |||
AK150677.1 | - | 3 | 445 | |||
AK166368.1 | - | 444 | ||||
AK167333.1 | - | 3 | 432 | |||
BC027437.1 | - | 3 | 434 | 4017054 | ||
CT010335.1 | - | 1 | 119 |