Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010137.3 | NP_034267.3 | 2 | 523 | ||
GenBank mRNA | AF045160.1 | - | 2 | 287 | ||
AK087208.1 | - | 1 | 1019 | |||
AK136911.1 | - | 2 | 524 | |||
AK141185.1 | - | 2 | 260 | |||
AK158430.1 | - | 2 | 687 | |||
BC057870.1 | - | 2 | 245 | 5032291 | ||
D89787.1 | - | 2 | 485 | |||
U81983.1 | - | 2 | 203 |