Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110228.1 | NP_001103698.1 | 2, 3 | 350 | ||
NM_001110229.1 | NP_001103699.1 | 3, 4 | 437 | |||
NM_001110230.1 | NP_001103700.1 | 3, 4 | 437 | |||
NM_001110231.1 | NP_001103701.1 | 5, 6 | 412 | |||
NM_001110232.1 | NP_001103702.1 | 4, 5 | 423 | |||
NM_001160292.1 | NP_001153764.1 | 3, 4 | 437 | |||
NM_001160293.1 | NP_001153765.1 | 2, 3 | 350 | |||
NM_010160.2 | NP_034290.2 | 2, 3 | 350 | |||
GenBank mRNA | AF090696.1 | - | 4, 5 | 423 | ||
AF090697.1 | - | 2, 3 | 323 | |||
AK037436.1 | - | 1, 2 | 809 | |||
AK135166.1 | - | 2, 3 | 301 | |||
AK137292.1 | - | 3, 4 | 378 | |||
AK137555.1 | - | 1, 2 | 208 | |||
AK151818.1 | - | 1, 2 | 208 | |||
AK152628.1 | - | 1, 2 | 208 | |||
AK153267.1 | - | 1, 2 | 208 | |||
AK159996.1 | - | 3, 4 | 444 | |||
AK160861.1 | - | 5, 6 | 413 | |||
AK162651.1 | - | 1, 2 | 208 | |||
AK164112.1 | - | 2, 3 | 247 | |||
AK164914.1 | - | 3, 4 | 378 | |||
AK169542.1 | - | 1, 2 | 223 | |||
AK172648.1 | - | 1, 2 | 208 | |||
AK206137.1 | - | 1, 2 | 36 | |||
BC026856.1 | - | 2, 3 | 301 | 4503295 | ||
BY145030.1 | - | 2, 3 | 307 | |||
CF536042.1 | - | 2, 3 | 350 | |||
Y18298.1 | - | 1, 2 | 216 |