Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eya1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252192.1 | NP_001239121.1 | 6, 7 | 1041 | ||
NM_010164.2 | NP_034294.2 | 7, 8 | 1157 | |||
GenBank mRNA | AK042412.1 | - | 6, 7 | 964 | ||
BC060260.1 | - | 7, 8 | 1157 | 6848408 | ||
BC066860.1 | - | 7, 8 | 811 | 30089692 | ||
U61110.1 | - | 6, 7 | 714 |