Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eci1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010023.4 | NP_034153.2 | 7 | 802 | ||
GenBank mRNA | AK004838.1 | - | 7 | 803 | ||
AK029481.1 | - | 7 | 803 | |||
AK077386.1 | - | 7 | 793 | |||
BC022712.1 | - | 7 | 786 | 4487371 | ||
BC054444.1 | - | 7 | 789 | 6476161 | ||
Z14049.1 | - | 7 | 782 |