Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010024.3 | NP_034154.2 | 1, 2 | 771 | ||
GenBank mRNA | AK033040.1 | - | 1, 2 | 770 | ||
AK212164.1 | - | 1, 2 | 349 | |||
AK214448.1 | - | 1, 2 | 330 | |||
AK217087.1 | - | 1, 2 | 331 | |||
AK219002.1 | - | 1, 2 | 358 | |||
BC067064.1 | - | 1, 2 | 605 | 30537997 | ||
BC082330.1 | - | 1, 2 | 406 | 30539879 | ||
EU554632.1 | - | 1, 2 | 303 | |||
X63349.1 | - | 1, 2 | 707 |