Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dio2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010050.2 | NP_034180.1 | 2 | 2096 | ||
GenBank mRNA | AF096875.1 | - | 2 | 2095 | ||
AK032038.1 | - | 2 | 1806 | |||
AK045607.1 | - | 2 | 1850 | |||
BC125383.1 | - | 2 | 1992 | 40129898 | ||
BC125385.1 | - | 2 | 1991 | 40129900 | ||
CB522312.1 | - | 1 | 426 |