Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010073.2 | NP_034203.1 | 3 | 171 | ||
GenBank mRNA | AB013360.1 | - | 3 | 143 | ||
AK002468.1 | - | 3 | 155 | |||
AK004241.1 | - | 3 | 146 | |||
BC008256.1 | - | 3 | 171 | 2648752 |