Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010074.3 | NP_034204.1 | 26 | 3271 | ||
GenBank mRNA | AK085370.1 | - | 26 | 2853 | ||
AK153939.1 | - | 26 | 2885 | |||
BC022183.1 | - | 27 | 3135 | 3982742 | ||
X58384.1 | - | 26 | 2949 |