Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276387.1 | NP_001263316.1 | 14, 15 | 2312 | ||
NM_010095.6 | NP_034225.1 | 14, 15 | 2307 | |||
GenBank mRNA | AK053013.1 | - | 14, 15 | 2308 | ||
AK139262.1 | - | 14, 15 | 2019 | |||
BC049188.1 | - | 13, 14 | 1606 | 6312260 | ||
BC050922.1 | - | 13, 14 | 1829 | 6409737 | ||
U71189.1 | - | 13, 14 | 1492 | |||
U82441.1 | - | 13, 14 | 1361 | |||
U92703.1 | - | 13, 14 | 1712 |