Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034996.1 | - | 5 | 2331 | ||
BC018572.1 | - | 5 | 2142 | |||
BC021656.1 | - | 5 | 2314 | 3602195 | ||
BQ177892.1 | - | 1 | 495 | |||
Z48781.1 | - | 5 | 2293 |