Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010119.5 | NP_034249.1 | 4 | 1035 | ||
GenBank mRNA | AF099186.1 | - | 4 | 1038 | ||
AF173156.1 | - | 4 | 1046 | |||
AK012896.1 | - | 4 | 1011 | |||
AK077536.1 | - | 4 | 1015 | |||
AK088980.1 | - | 5 | 1177 | |||
AK152821.1 | - | 4 | 984 | |||
AK154651.1 | - | 4 | 1024 | |||
AK159191.1 | - | 4 | 1010 | |||
AK159943.1 | - | 4 | 1020 | |||
AK160535.1 | - | 4 | 1023 | |||
AK166701.1 | - | 4 | 1021 | |||
AK167721.1 | - | 4 | 1027 | |||
AK168610.1 | - | 4 | 1011 | |||
AK170679.1 | - | 4 | 811 | |||
AK171047.1 | - | 4 | 1039 | |||
AK172539.1 | - | 4 | 1011 | |||
AK172669.1 | - | 4 | 1197 | |||
BC037094.1 | - | 4 | 1022 | |||
BC043332.1 | - | 4 | 1005 | 4981832 | ||
BC054828.1 | - | 4 | 999 | 5042951 |