Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159571.1 | NP_001153043.1 | 11 | 2288 | ||
NM_010144.6 | NP_034274.4 | 11 | 2261 | |||
GenBank mRNA | AK046884.1 | - | 11 | 2226 | ||
AK049340.1 | - | 11 | 2262 | |||
AK132392.1 | - | 11 | 2261 | |||
AK143422.1 | - | 2231 | ||||
BC013342.1 | - | 11 | 1949 | |||
BC016505.1 | - | 11 | 2213 | 3709926 | ||
BC090839.1 | - | 11 | 2038 | |||
U06834.1 | - | 12 | 1874 |