Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110228.1 | NP_001103698.1 | 14 | 2134 | ||
NM_001110229.1 | NP_001103699.1 | 15 | 2233 | |||
NM_001110230.1 | NP_001103700.1 | 15 | 2221 | |||
NM_001110231.1 | NP_001103701.1 | 16 | 3580 | |||
NM_001110232.1 | NP_001103702.1 | 16 | 2207 | |||
NM_001160292.1 | NP_001153764.1 | 14 | 2077 | |||
NM_001160293.1 | NP_001153765.1 | 14 | 3238 | |||
NM_010160.2 | NP_034290.2 | 12 | 3356 | |||
GenBank mRNA | AF090696.1 | - | 16 | 2207 | ||
AF090697.1 | - | 14 | 2107 | |||
AK152628.1 | - | 12 | 1860 | |||
AK159996.1 | - | 14 | 2080 | |||
AK164914.1 | - | 15 | 2162 | |||
BC026856.1 | - | 14 | 3189 | 4503295 |