Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fasl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205243.1 | NP_001192172.1 | 1 | 297 | ||
NM_010177.4 | NP_034307.1 | 1 | 297 | |||
GenBank mRNA | AF288572.1 | - | 1 | 87 | ||
AK040139.1 | - | 1 | 297 | |||
AK141268.1 | - | 1 | 297 | |||
BC052866.1 | - | 394 | 30073084 | |||
DQ846747.1 | - | 1 | 87 | |||
S76752.1 | - | 1 | 117 | |||
U06948.1 | - | 1 | 211 | |||
U10984.1 | - | 1 | 267 | |||
U58995.1 | - | 1 | 87 |