Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC34A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177316.1 | NP_001170787.1 | 10 | 1305 | ||
NM_001177317.1 | NP_001170788.1 | 10 | 1308 | |||
NM_080877.2 | NP_543153.1 | 10 | 1270 | |||
XM_011518257.2 | XP_011516559.1 | 10 | 1246 | |||
XM_011518258.1 | XP_011516560.1 | 9 | 1136 | |||
XM_011518259.1 | XP_011516561.1 | 10 | 1270 | |||
XM_011518260.1 | XP_011516562.1 | 10 | 1188 | |||
XM_011518261.2 | XP_011516563.1 | 10 | 1261 | |||
XM_017014290.1 | XP_016869779.1 | 11 | 1356 | |||
XM_017014291.1 | XP_016869780.1 | 10 | 1246 | |||
XM_017014292.1 | XP_016869781.1 | 10 | 1246 | |||
GenBank mRNA | AB055000.1 | - | 10 | 1154 | ||
AK095999.1 | - | 10 | 1270 |