Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010201.4 | NP_034331.2 | 2 | 240 | ||
NM_207667.3 | NP_997550.1 | 2 | 640 | |||
GenBank mRNA | AB008907.1 | - | 2 | 230 | ||
AK030616.1 | - | 2 | 599 | |||
AK035974.1 | - | 2 | 250 | |||
AK079100.1 | - | 2 | 640 | |||
BC107003.1 | - | 3 | 574 | 40046696 | ||
BC107004.1 | - | 2 | 534 | 40046697 | ||
U66204.1 | - | 2 | 215 |