Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010219.3 | NP_034349.1 | 1, 2 | 234 | ||
GenBank mRNA | AK051433.1 | - | 260 | |||
AK083912.1 | - | 1, 2 | 234 | |||
AK089972.1 | - | 1, 2 | 245 | |||
AK159211.1 | - | 1, 2 | 260 | |||
AK160202.1 | - | 1, 2 | 183 | |||
AK212190.1 | - | 1, 2 | 88 | |||
BC003447.1 | - | 1, 2 | 251 | 2651490 | ||
X17069.2 | - | 1, 2 | 100 | |||
X70887.1 | - | 1, 2 | 131 |