Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163481.1 | NP_001156953.1 | 4 | 690 | ||
NM_010221.2 | NP_034351.2 | 4 | 690 | |||
GenBank mRNA | AK019446.2 | - | 4 | 628 | ||
AK144185.1 | - | 4 | 673 | |||
BC029546.1 | - | 4 | 637 | 5059670 | ||
L07063.1 | - | 4 | 693 |