Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133267.2 | NP_573574.1 | 2 | 967 | ||
GenBank mRNA | AB028838.1 | - | 2 | 1017 | ||
AI360912.1 | - | 2 | 232 | |||
BC075089.2 | - | 2 | 725 | 30915389 | ||
BC075090.2 | - | 2 | 725 | 30915601 |