Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010242.3 | NP_034372.1 | 1 | 715 | ||
GenBank mRNA | AK041752.1 | - | 1 | 715 | ||
BC137588.1 | - | 1 | 706 | 8860607 |