Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010250.5 | NP_034380.1 | 3, 4 | 963 | ||
GenBank mRNA | AK034095.1 | - | 3, 4 | 964 | ||
AK039496.1 | - | 3, 4 | 247 | |||
AK141596.1 | - | 3, 4 | 823 | |||
BC132331.1 | - | 3, 4 | 639 | 40130608 | ||
BC137733.1 | - | 3, 4 | 642 | 8860752 | ||
M63436.1 | - | 3, 4 | 636 | |||
M86566.1 | - | 3, 4 | 331 | |||
X61430.1 | - | 2, 3 | 219 |