Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010264.4 | NP_034394.1 | 2 | 363 | ||
GenBank mRNA | AF390896.1 | - | 3 | 508 | ||
CN538225.1 | - | 1 | 34 | |||
U09563.1 | - | 2 | 363 | |||
U14666.1 | - | 2 | 322 |