Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145820.1 | NP_001139292.1 | 3, 4 | 601 | ||
NM_010274.3 | NP_034404.3 | 3, 4 | 476 | |||
GenBank mRNA | AK034353.1 | - | 3, 4 | 385 | ||
AK144716.1 | - | 3, 4 | 602 | |||
AK151189.1 | - | 3, 4 | 388 | |||
AK162556.1 | - | 3, 4 | 364 | |||
AK167152.1 | - | 3, 4 | 414 | |||
BC021359.1 | - | 3, 4 | 570 | 5012142 | ||
D50430.1 | - | 3, 4 | 378 | |||
U60987.1 | - | 3, 4 | 412 |