Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gem | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010276.4 | NP_034406.2 | 2 | 220 | ||
GenBank mRNA | AK153999.1 | - | 1 | 1551 | ||
AK157655.1 | - | 2 | 222 | |||
AK158393.1 | - | 2 | 222 | |||
AK168401.1 | - | 2 | 221 | |||
AK169699.1 | - | 1 | 1551 | |||
AK171336.1 | - | 2 | 158 | |||
BC029668.1 | - | 2 | 196 | 5368352 | ||
BY090781.1 | - | 2 | 268 | |||
BY133754.1 | - | 2 | 283 | |||
U10551.1 | - | 1 | 199 | |||
U13053.1 | - | 2 | 211 |