Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267621.1 | NP_001254550.1 | 5, 6 | 1228 | ||
NM_010278.2 | NP_034408.1 | 4, 5 | 1440 | |||
GenBank mRNA | AK089065.1 | - | 5, 6 | 1229 | ||
BC130219.1 | - | 3, 4 | 625 | |||
BC152328.1 | - | 3, 4 | 516 | |||
BU698904.1 | - | 2, 3 | 256 | |||
U58972.1 | - | 4, 5 | 1415 | |||
U78312.1 | - | 4, 5 | 1440 |