Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcer1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010184.2 | NP_034314.2 | 3 | 275 | ||
GenBank mRNA | BC125455.1 | - | 3 | 240 | 40129970 | |
BC125457.1 | - | 3 | 239 | 40129972 | ||
BC145138.1 | - | 3 | 197 | |||
CJ142154.1 | - | 3 | 196 | |||
CJ142191.1 | - | 3 | 275 | |||
J05018.1 | - | 3 | 260 |