Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010198.2 | NP_034328.1 | 4 | 537 | ||
GenBank mRNA | U66203.1 | - | 4 | 446 |