Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF047699.1 | - | 8 | 832 | ||
AF055573.1 | - | 698 | ||||
BC012662.1 | - | 6 | 557 | 4036815 |