Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111066.1 | NP_001104536.1 | 6 | 911 | ||
NM_001199631.1 | NP_001186560.1 | 6 | 1007 | |||
NM_010223.2 | NP_034353.2 | 6 | 908 | |||
GenBank mRNA | AB154247.1 | - | 5 | 647 | ||
AF030635.1 | - | 5 | 678 | |||
AK088739.1 | - | 6 | 846 | |||
AK146122.1 | - | 6 | 906 | |||
AK152198.1 | - | 6 | 907 | |||
AK153000.1 | - | 6 | 904 | |||
AK165281.1 | - | 6 | 904 | |||
AK167663.1 | - | 6 | 894 | |||
AK177591.1 | - | 2 | 102 | |||
AK179469.1 | - | 2 | 155 | |||
AK206245.1 | - | 2 | 102 | |||
AK209719.1 | - | 3 | 311 | |||
AK212094.1 | - | 3 | 309 | |||
AY225340.1 | - | 5 | 791 | |||
AY278608.1 | - | 6 | 867 | |||
BC003739.1 | - | 5 | 827 | 3601511 | ||
BC027808.1 | - | 6 | 861 | 5324380 |