Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285458.1 | NP_001272387.1 | 2 | 330 | ||
NM_010230.2 | NP_034360.2 | 2 | 330 | |||
GenBank mRNA | AK135758.1 | - | 2 | 296 | ||
BC138036.1 | - | 1 | 230 | 8861055 | ||
BC171945.1 | - | 1 | 230 | 9054231 | ||
BY288387.1 | - | 2 | 331 | |||
X53599.1 | - | 2 | 284 |