Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010231.2 | NP_034361.1 | 4, 5 | 808 | ||
GenBank mRNA | AK143940.1 | - | 4, 5 | 638 | ||
BC011229.1 | - | 4, 5 | 808 | 4196446 | ||
D16215.1 | - | 4, 5 | 698 | |||
U87456.1 | - | 4, 5 | 793 |