Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gamt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010255.3 | NP_034385.1 | 2, 3 | 402 | ||
GenBank mRNA | AA881765.1 | - | 2, 3 | 418 | ||
AF015887.1 | - | 2, 3 | 335 | |||
AK140688.1 | - | 2, 3 | 403 | |||
AK158087.1 | - | 2, 3 | 345 | |||
BC049233.1 | - | 2, 3 | 388 | 4195030 |