Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbp2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010259.2 | NP_034389.2 | 5, 6 | 712 | ||
GenBank mRNA | AK077796.1 | - | 5, 6 | 713 | ||
BC108990.2 | - | 757 | 40047769 | |||
EF494422.1 | - | 5, 6 | 712 | |||
M55544.1 | - | 696 | ||||
M63961.1 | - | 790 |