Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010267.3 | NP_034397.1 | 1, 2 | 224 | ||
GenBank mRNA | AK020988.1 | - | 1, 2 | 208 | ||
AK045460.1 | - | 1, 2 | 224 | |||
AK049655.1 | - | 1, 2 | 194 | |||
AK083814.1 | - | 1, 2 | 225 | |||
AK139057.1 | - | 1, 2 | 67 | |||
AK149281.1 | - | 1, 2 | 235 | |||
BC048177.1 | - | 1, 2 | 200 | 6825756 | ||
BC051135.1 | - | 1, 2 | 189 | 6532839 | ||
BC060212.1 | - | 1, 2 | 197 | |||
Y17850.1 | - | 1, 2 | 212 |