Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010269.3 | NP_034399.1 | 3 | 569 | ||
GenBank mRNA | AK004887.1 | - | 3 | 515 | ||
AK082958.1 | - | 3 | 531 | |||
AK146835.1 | - | 3 | 559 | |||
AK161122.1 | - | 3 | 506 | |||
AK166799.1 | - | 3 | 484 | |||
AK166859.1 | - | 3 | 483 | |||
BC025070.1 | - | 3 | 478 | 3710712 | ||
Y17851.1 | - | 3 | 567 |