Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145820.1 | NP_001139292.1 | 4 | 699 | ||
NM_010274.3 | NP_034404.3 | 4 | 574 | |||
GenBank mRNA | AK034353.1 | - | 4 | 483 | ||
AK144716.1 | - | 4 | 700 | |||
AK151189.1 | - | 4 | 486 | |||
AK162556.1 | - | 4 | 462 | |||
AK167152.1 | - | 4 | 512 | |||
BC021359.1 | - | 4 | 668 | 5012142 | ||
D50430.1 | - | 4 | 476 | |||
U60987.1 | - | 4 | 510 |