Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eya4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010167.4 | NP_034297.2 | 13, 14 | 1428 | ||
GenBank mRNA | AK041625.1 | - | 13, 14 | 1429 | ||
BC120898.1 | - | 13, 14 | 1412 | 40061597 | ||
BC120899.1 | - | 13, 14 | 1412 | 40061599 | ||
Y17115.1 | - | 14, 15 | 1539 |