Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fadd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010175.5 | NP_034305.1 | 2 | 2179 | ||
GenBank mRNA | AK084808.1 | - | 2 | 2180 | ||
AK169798.1 | - | 2 | 2208 |