Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284196.1 | NP_001271125.1 | 6 | 1078 | ||
NM_010178.3 | NP_034308.1 | 6 | 1081 | |||
GenBank mRNA | AB015895.1 | - | 6 | 989 | ||
AF042383.1 | - | 6 | 993 | |||
AK030968.1 | - | 1 | 1096 | |||
AK149716.1 | - | 6 | 1008 | |||
AK150072.1 | - | 6 | 1008 | |||
AK150734.1 | - | 6 | 1015 | |||
AK151567.1 | - | 6 | 1020 | |||
AK168571.1 | - | 6 | 1004 | |||
AK169071.1 | - | 6 | 1005 |