Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003487.3 | NP_003478.1 | 3 | 202 | ||
NM_139215.2 | NP_631961.1 | 3 | 202 | |||
XM_011525315.1 | XP_011523617.1 | 3 | 172 | |||
XM_017025181.1 | XP_016880670.1 | 2 | 80 | |||
GenBank mRNA | AK304520.1 | - | 3 | 173 | ||
AK307637.1 | - | 3 | 213 | |||
AK313223.1 | - | 3 | 168 | |||
AK314194.1 | - | 3 | 173 | |||
BC046099.1 | - | 3 | 134 | 6067044 | ||
DB166653.1 | - | 3 | 202 | |||
U51334.1 | - | 3 | 173 | |||
X98893.1 | - | 3 | 133 |