Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fdft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076062.1 | - | 7 | 1012 | ||
AK136159.1 | - | 9 | 1794 | |||
AK146766.1 | - | 7 | 1020 | |||
AK158633.1 | - | 7 | 1009 | |||
AK162294.1 | - | 7 | 973 | |||
AK168629.1 | - | 7 | 946 | |||
AK169317.1 | - | 7 | 1025 | |||
AK183967.1 | - | 2 | 102 | |||
AK217297.1 | - | 2 | 102 | |||
BC054722.1 | - | 7 | 954 | 5694816 | ||
BC138301.1 | - | 7 | 1008 | 8861320 | ||
BC138302.1 | - | 7 | 1009 | 8861321 | ||
D29016.1 | - | 7 | 962 |