Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111066.1 | NP_001104536.1 | 9 | 1444 | ||
NM_001199631.1 | NP_001186560.1 | 9 | 1540 | |||
NM_010223.2 | NP_034353.2 | 9 | 1441 | |||
GenBank mRNA | AF030635.1 | - | 8 | 1211 | ||
AK088739.1 | - | 9 | 1379 | |||
AK146122.1 | - | 9 | 1439 | |||
AK152198.1 | - | 9 | 1440 | |||
AK153000.1 | - | 9 | 1433 | |||
AK165281.1 | - | 9 | 1437 | |||
AK167663.1 | - | 9 | 1427 | |||
AK178628.1 | - | 1 | 169 | |||
AK188492.1 | - | 1 | 164 | |||
AK189244.1 | - | 1 | 172 | |||
AK190969.1 | - | 1 | 143 | |||
AK192821.1 | - | 1 | 117 | |||
AK205574.1 | - | 1 | 139 | |||
AK207414.1 | - | 1 | 123 | |||
AK208969.1 | - | 1 | 111 | |||
AK212129.1 | - | 1 | 138 | |||
AK213138.1 | - | 1 | 147 | |||
AK214360.1 | - | 1 | 128 | |||
AK218022.1 | - | 2 | 219 | |||
AY278608.1 | - | 9 | 1400 | |||
BC003739.1 | - | 8 | 1360 | 3601511 | ||
BC027808.1 | - | 9 | 1394 | 5324380 |