Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010229.2 | NP_034359.2 | 12 | 1676 | ||
GenBank mRNA | AK149292.1 | - | 12 | 1494 | ||
AK163640.1 | - | 12 | 1677 | |||
BC109003.1 | - | 12 | 1469 | |||
BC109004.1 | - | 12 | 1481 | |||
M64689.1 | - | 12 | 1482 | |||
X59398.1 | - | 12 | 1533 |