Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fth1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010239.2 | NP_034369.1 | 4 | 840 | ||
GenBank mRNA | AK027998.1 | - | 4 | 783 | ||
AK088935.1 | - | 3 | 1988 | |||
AK139622.1 | - | 4 | 783 | |||
AK147082.1 | - | 4 | 786 | |||
AK150262.1 | - | 4 | 783 | |||
AK150508.1 | - | 4 | 783 | |||
AK150628.1 | - | 4 | 783 | |||
AK150679.1 | - | 4 | 786 | |||
AK150693.1 | - | 4 | 783 | |||
AK151192.1 | - | 4 | 783 | |||
AK151241.1 | - | 4 | 786 | |||
AK151399.1 | - | 4 | 769 | |||
AK151609.1 | - | 4 | 783 | |||
AK151675.1 | - | 4 | 783 | |||
AK152071.1 | - | 4 | 783 | |||
AK152542.1 | - | 4 | 786 | |||
AK152702.1 | - | 5 | 1411 | |||
AK153017.1 | - | 4 | 783 | |||
AK153195.1 | - | 4 | 786 | |||
AK153199.1 | - | 4 | 786 | |||
AK159243.1 | - | 4 | 783 | |||
AK168601.1 | - | 4 | 783 | |||
AK169004.1 | - | 4 | 783 | |||
AK214283.1 | - | 1 | 48 | |||
BC011096.1 | - | 4 | 766 | |||
BC012314.1 | - | 4 | 761 | 3488821 | ||
J03941.1 | - | 4 | 736 | |||
X12812.1 | - | 4 | 682 |