Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gamt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010255.3 | NP_034385.1 | 6 | 669 | ||
GenBank mRNA | AF015887.1 | - | 6 | 602 | ||
AK140688.1 | - | 6 | 718 | |||
AK158087.1 | - | 6 | 612 | |||
AK178068.1 | - | 2 | 88 | |||
AK217844.1 | - | 2 | 89 | |||
BC049233.1 | - | 6 | 655 | 4195030 |