Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159548.1 | NP_001153020.1 | 9 | 4790 | ||
NM_001159549.1 | NP_001153021.1 | 9 | 4787 | |||
NM_010264.4 | NP_034394.1 | 11 | 5098 | |||
GenBank mRNA | AF390896.1 | - | 12 | 5236 | ||
AK164983.1 | - | 1 | 349 |