Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010266.2 | NP_034396.1 | 11 | 1250 | ||
GenBank mRNA | AF174583.1 | - | 11 | 1176 | ||
AF369880.1 | - | 11 | 1135 | |||
AF369881.1 | - | 11 | 1135 | |||
AF369882.1 | - | 11 | 1135 | |||
AK041056.1 | - | 11 | 1189 | |||
AK079419.1 | - | 11 | 1058 | |||
AK079459.1 | - | 11 | 1189 | |||
AK139617.1 | - | 1019 | ||||
AK157014.1 | - | 11 | 1212 | |||
AK173138.1 | - | 11 | 1179 | |||
BC076583.1 | - | 11 | 1163 | 30620457 |