Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010287.2 | NP_034417.1 | 2 | 711 | ||
GenBank mRNA | AK035356.1 | - | 2 | 756 | ||
AK081276.1 | - | 2 | 724 | |||
M80481.1 | - | 2 | 712 |
Set Membership: |
G-protein coupled receptor |