Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gja1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010288.3 | NP_034418.1 | 2 | 546 | ||
GenBank mRNA | AK011459.1 | - | 2 | 527 | ||
AK029291.1 | - | 2 | 403 | |||
AK036979.1 | - | 2 | 517 | |||
AK145692.1 | - | 2 | 516 | |||
AK165986.1 | - | 2 | 403 | |||
AK171955.1 | - | 2 | 515 | |||
BC006894.1 | - | 2 | 499 | 3599347 | ||
BC011324.1 | - | 2 | 627 | |||
BC055375.1 | - | 2 | 469 | 30063597 | ||
CT010327.1 | - | 1 | 314 | |||
M63801.1 | - | 2 | 534 | |||
X61576.1 | - | 2 | 400 |